3-Bromo-4-methylbenzaldehyde ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
B5073-1G
B5073-5G
TCB5073-1GEA
36
USD
TCB5073-1G
TCB5073-5G
3-Bromo-4-methylbenzaldehyde ≥96.0% (by GC)
3-Bromo-4-methylbenzaldehyde
Formula:
C₈H₇BrO |
MDL Number:
MFCD04971197 CAS Number: 36276-24-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...