Ethyl-3-hydroxy-2-naphthoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Hydroxy-2-naphthoic acid ethyl ester, Ethyl 3-hydroxy-2-naphthoate

E1249-1G E1249-5G
TCE1249-1GEA 52 USD
TCE1249-1G TCE1249-5G
Ethyl-3-hydroxy-2-naphthoate ≥98.0% (by GC)
Ethyl 3-hydroxy-2-naphthoate
Formula: C₁₃H₁₂O₃
MW: 216.2326 g/mol
Boiling Pt: 291°C
MDL Number: MFCD00220608
CAS Number: 7163-25-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR