5-Acetylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Acetylthiophene-2-carboxylic Acid, 5-Acetyl-2-thenoic acid

A2866-1G A2866-5G
TCA2866-1GEA 28 USD
TCA2866-1G TCA2866-5G
5-Acetylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Acetylthiophene-2-carboxylic acid
Formula: C₇H₆O₃S
MW: 170.19 g/mol
MDL Number: MFCD00055512
CAS Number: 4066-41-5

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR