[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: [2-(2-Methoxyethoxy)ethoxy]acetic acid, 3,6,9-Trioxadecanoic acid, 2-(2-(2-Methoxyethoxy)ethoxy)acetic acid

M2963-25G M2963-5G
76220-312EA 154 USD
76220-312 76220-314
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) ≥95.0% (by GC, titration analysis)
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid)
Formula: C₇H₁₄O₅
MW: 178.1831 g/mol
Boiling Pt: 107°C
MDL Number: MFCD00044098
CAS Number: 16024-58-1
EINECS: 240-162-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR