Methyl oxindole-6-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 2-Oxoindoline-6-carboxylate, 2-Oxoindoline-6-carboxylic acid methyll ester

M2799-1G M2799-5G
TCM2799-1GEA 39 USD
TCM2799-1G TCM2799-5G
Methyl oxindole-6-carboxylate ≥98.0% (by GC)
Methyl oxindole-6-carboxylate
Formula: C₁₀H₉NO₃
MW: 191.1800 g/mol
MDL Number: MFCD03095196
CAS Number: 14192-26-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 210.0 to 214.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR