4-Bromotetrahydropyran ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Bromooxane, 4-Bromotetrahydro-2H-pyran

B5600-1G B5600-5G
TCB5600-1GEA 69 USD
TCB5600-1G TCB5600-5G
4-Bromotetrahydropyran ≥98.0% (by GC)
4-Bromotetrahydropyran
Formula: C₅H₉BrO
MW: 165.0300 g/mol
Boiling Pt: 61°C
Flash Pt: 71°C
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now

Specification Test Results

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR