4-Phosphonobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

P2349-1G P2349-200MG
TCP2349-1GEA 274 USD
TCP2349-1G TCP2349-200MG
4-Phosphonobutyric acid ≥98.0% (by titrimetric analysis)
4-Phosphonobutyric acid
Formula: C₄H₉O₅P
MDL Number: MFCD00041414
CAS Number: 4378-43-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR