4-Chloro-1-iodo-2-nitrobenzene ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C2932-1G
C2932-5G
TCC2932-1GEA
65
USD
TCC2932-1G
TCC2932-5G
4-Chloro-1-iodo-2-nitrobenzene ≥98.0% (by GC)
4-Chloro-1-iodo-2-nitrobenzene
Formula:
C₆H₃ClINO₂ |
MDL Number:
MFCD11226294 CAS Number: 5446-05-9 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...