Creatine (N-amidinosarcosine) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(1-Methylguanidino)acetic acid, N-Amidinosarcosine, Creatine

C3610-100G C3610-25G
TCC3610-100GEA 53 USD
TCC3610-100G TCC3610-25G
Creatine (N-amidinosarcosine) ≥98.0% (by HPLC, titration analysis)
Creatine (N-amidinosarcosine)
Formula: C₄H₉N₃O₂
Melting Pt: 303°C
MDL Number: MFCD00004282
CAS Number: 57-00-1
EINECS: 200-306-6
Merck Index: 14,02568

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR