3-Bromo-4-fluorophenol ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4491-25G B4491-5G
TCB4491-25GEA 168 USD
TCB4491-25G TCB4491-5G
3-Bromo-4-fluorophenol ≥99.0% (by GC, titration analysis)
3-Bromo-4-fluorophenol
Formula: C₆H₄BrFO
Boiling Pt: 78°C
Storage Temperature: Refrigerator
MDL Number: MFCD03425884
CAS Number: 27407-11-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR