Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
T3623-25G
T3623-5G
TCT3623-25GEA
146
USD
TCT3623-25G
TCT3623-5G
Trihexyl(tetradecyl)phosphonium chloride ≥97.0% (by titrimetric analysis)
Trihexyl(tetradecyl)phosphonium chloride
Formula:
C₃₂H₆₈ClP MW: 519.31 g/mol |
MDL Number:
MFCD04112746 CAS Number: 258864-54-9 UN: 1760 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...