2-Bromo-4-fluoropyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4760-1G B4760-5G
TCB4760-1GEA 50 USD
TCB4760-1G TCB4760-5G
2-Bromo-4-fluoropyridine ≥98.0% (by GC)
2-Bromo-4-fluoropyridine
Formula: C₅H₃BrFN
Flash Pt: 76°C
MDL Number: MFCD09260909
CAS Number: 357927-50-5

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR