2-(Trifluoromethyl)nicotinic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Trifluoromethylnicotinic acid

T3132-1G T3132-5G
TCT3132-1GEA 92 USD
TCT3132-1G TCT3132-5G
2-(Trifluoromethyl)nicotinic acid ≥97.0% (by GC, titration analysis)
2-(Trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₂
MDL Number: MFCD01862052
CAS Number: 131747-43-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 185.0 to 189.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR