Octylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

O0380-1G O0380-5G
TCO0380-1GEA 64 USD
TCO0380-1G TCO0380-5G
Octylphosphonic acid ≥98.0% (by titrimetric analysis)
Octylphosphonic acid
Formula: C₈H₁₉O₃P
MDL Number: MFCD00015841
CAS Number: 4724-48-5
EINECS: 225-218-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR