3-Bromobenzo[b]thiophene ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
B5589-25G
B5589-5G
TCB5589-25GEA
212
USD
TCB5589-25G
TCB5589-5G
3-Bromobenzo[b]thiophene ≥96.0% (by GC)
3-Bromobenzo[b]thiophene
Formula:
C₈H₅BrS Boiling Pt: 269°C |
MDL Number:
MFCD00023009 CAS Number: 7342-82-7 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...