3,4,5-Tribromopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3248-1G T3248-200MG
TCT3248-1GEA 142.3 USD
TCT3248-1G TCT3248-200MG
3,4,5-Tribromopyridine ≥98.0% (by GC)
3,4,5-Tribromopyridine
Formula: C₅H₂Br₃N
MW: 315.79 g/mol
Boiling Pt: 285°C
Melting Pt: 107°C
MDL Number: MFCD01646053
CAS Number: 2457-48-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR