7-Bromoisoquinoline ≥96.0% (by GC)

Supplier: TCI America
Warning

B5071-1G B5071-200MG
TCB5071-1GEA 215 USD
TCB5071-1G TCB5071-200MG
7-Bromoisoquinoline ≥96.0% (by GC)
7-Bromoisoquinoline
Formula: C₉H₆BrN
MDL Number: MFCD07368661
CAS Number: 58794-09-5

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR