3-Chloro-4-methylanisole ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Chloro-4-methoxy-1-methylbenzene, 3-Chloro-4-methylanisole, 2-Chloro-4-methoxytoluene
C3190-25G
C3190-5G
TCC3190-25GEA
322
USD
TCC3190-25G
TCC3190-5G
3-Chloro-4-methylanisole ≥95.0% (by GC)
3-Chloro-4-methylanisole
Formula:
C₈H₉ClO MW: 156.61 g/mol Boiling Pt: 213°C |
MDL Number:
MFCD00087376 CAS Number: 54788-38-4 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...