4-tert-Butylcalix[6]arene
Supplier: TCI America
|
Warning
|
Synonyms:
p-tert-Butylcalix[6]arene
B5375-1G
B5375-5G
TCB5375-1GEA
36
USD
TCB5375-1G
TCB5375-5G
4-tert-Butylcalix[6]arene
4-tert-Butylcalix[6]arene
Formula:
C₆₆H₈₄O₆ MW: 973.39 g/mol Melting Pt: 381°C |
MDL Number:
MFCD00075464 CAS Number: 78092-53-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% (Except for the toluene) |
Residual Solvent | max. 10.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...