2-Bromo-1-tetralone ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-3,4-dihydronaphthalen-1(2H)-one, 2-Bromo-3,4-dihydro-1(2H)-naphthalenone

B5698-1G B5698-5G
TCB5698-1GEA 73 USD
TCB5698-1G TCB5698-5G
2-Bromo-1-tetralone ≥97.0% (by GC)
2-Bromo-1-tetralone
Formula: C₁₀H₉BrO
MW: 225.08 g/mol
MDL Number: MFCD00089999
CAS Number: 13672-07-6
EINECS: 237-154-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR