3-Aminocoumarin ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Amino-2H-chromen-2-one
A2816-1G
A2816-200MG
TCA2816-1GEA
162
USD
TCA2816-1G
TCA2816-200MG
3-Aminocoumarin ≥98.0% (by GC)
3-Aminocoumarin
Formula:
C₉H₇NO₂ MW: 161.16 g/mol |
MDL Number:
MFCD00016965 CAS Number: 1635-31-0 EINECS: 216-659-4 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...