9-Aminoacridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 9-AA

A2905-1G A2905-5G
TCA2905-1GEA 58 USD
TCA2905-1G TCA2905-5G
9-Aminoacridine ≥97.0% (by GC, titration analysis)
9-Aminoacridine
Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
MDL Number: MFCD00037839
CAS Number: 90-45-9
EINECS: 201-995-6
Merck Index: 14,00407

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 236.0 to 240.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR