1-Butyl-1-methylpyrrolidiniumhexafluorophosphate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: N-Butyl-N-methylpyrrolidinium hexafluorophosphate

B6039-25G B6039-5G
TCB6039-25GEA 201 USD
TCB6039-25G TCB6039-5G
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate ≥98.0% (by total nitrogen basis)
1-Butyl-1-methylpyrrolidiniumhexafluorophosphate
Formula: C₉H₂₀F₆NP
MDL Number: MFCD08458501
CAS Number: 330671-29-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 84.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR