Dimethoxy(diisobutyl)silane ≥98.0% (by GC)

Supplier: TCI America
Warning

D4727-25ML D4727-5ML
TCD4727-25MLEA 52 USD
TCD4727-25ML TCD4727-5ML
Dimethoxy(diisobutyl)silane ≥98.0% (by GC)
Dimethoxy(diisobutyl)silane
Formula: C₁₀H₂₄O₂Si
Boiling Pt: 72°C
Flash Pt: 102°C
MDL Number: MFCD00236510
CAS Number: 17980-32-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR