5-Nitropicolinic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid
N0930-1G
N0930-5G
TCN0930-1GEA
86.8
USD
TCN0930-1G
TCN0930-5G
5-Nitropicolinic acid ≥98.0% (by HPLC, titration analysis)
5-Nitropicolinic acid
Formula:
C₆H₄N₂O₄ MW: 168.11 g/mol |
MDL Number:
MFCD04114182 CAS Number: 30651-24-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...