Methyl-2,5-dichloronicotinate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Methyl 2,5-dichloronicotinate
M2752-1G
M2752-200MG
TCM2752-1GEA
100.4
USD
TCM2752-1G
TCM2752-200MG
Methyl-2,5-dichloronicotinate ≥98.0% (by GC)
Methyl-2,5-dichloronicotinate
Formula:
C₇H₅Cl₂NO₂ MW: 206.03 g/mol |
MDL Number:
MFCD03844857 CAS Number: 67754-03-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...