2-Acetamido-5-bromobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Acetamido-5-bromobenzoic Acid, N-Acetyl-5-bromoanthranilic acid
A2839-1G
A2839-5G
TCA2839-1GEA
33
USD
TCA2839-1G
TCA2839-5G
2-Acetamido-5-bromobenzoic acid ≥98.0% (by GC, titration analysis)
2-Acetamido-5-bromobenzoic acid
Formula:
C₉H₈BrNO₃ |
MDL Number:
MFCD00040904 CAS Number: 38985-79-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...