Hydrazinium monobromide ∼50% in water
Supplier: TCI America
|
Danger
|
Synonyms:
Hydrazine monohydrobromide
H1517-25G
H1517-500G
TCH1517-25GEA
32
USD
TCH1517-25G
TCH1517-500G
Hydrazinium monobromide ∼50% in water
Hydrazinium monobromide
Formula:
H₅BrN₂ MW: 112.96 g/mol |
MDL Number:
MFCD00065394 CAS Number: 13775-80-9 EINECS: 237-412-7 UN: 3289 ADR: 6.1,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Potassium iodate Method) | 49.0 to 51.0 w/w% |
Concentration (Argentmetric Titration) | 49.0 to 51.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...