Methyl benzenesulfinate ≥98.0% (by GC)
Supplier: TCI America
M2971-25G
M2971-5G
TCM2971-25GEA
453
USD
TCM2971-25G
TCM2971-5G
Methyl benzenesulfinate ≥98.0% (by GC)
Methyl benzenesulphinate
Formula:
C₇H₈O₂S Boiling Pt: 63°C Flash Pt: 92°C |
MDL Number:
MFCD00674051 CAS Number: 670-98-4 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...