2-Acetamido-6-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

A2780-25G A2780-5G
TCA2780-25GEA 303 USD
TCA2780-25G TCA2780-5G
2-Acetamido-6-methylpyridine ≥98.0% (by GC)
2-Acetamido-6-methylpyridine
Formula: C₈H₁₀N₂O
MDL Number: MFCD00091881
CAS Number: 5327-33-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR