Barium trifluoromethanesulphonate ≥96.0% (W)

Supplier: TCI America
Danger

Synonyms: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

B6120-25G B6120-5G
TCB6120-25GEA 146 USD
TCB6120-25G TCB6120-5G
Barium trifluoromethanesulphonate ≥96.0% (W)
Barium trifluoromethanesulphonate
Formula: C₂BaF₆O₆S₂
MW: 435.45 g/mol
Melting Pt: 370°C
MDL Number: MFCD00143644
CAS Number: 2794-60-7
EINECS: 220-526-6
UN: 1564
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR