Methyl-5-bromo-2-furoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2538-1G M2538-5G
TCM2538-1GEA 43.9 USD
TCM2538-1G TCM2538-5G
Methyl-5-bromo-2-furoate ≥98.0% (by GC)
Methyl-5-bromo-2-furoate
Formula: C₆H₅BrO₃
Boiling Pt: 80°C
MDL Number: MFCD00092313
CAS Number: 2527-99-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR