2-Amino-5-iodo-3-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2683-1G A2683-5G
TCA2683-1GEA 97 USD
TCA2683-1G TCA2683-5G
2-Amino-5-iodo-3-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-iodo-3-methylpyridine
Formula: C₆H₇IN₂
MDL Number: MFCD02102422
CAS Number: 166266-19-9

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR