4-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
M2840-1G
M2840-5G
TCM2840-1GEA
69
USD
TCM2840-1G
TCM2840-5G
4-Methyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
4-Methyl-2-thiopheneboronic acid
Formula:
C₅H₇BO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD01319040 CAS Number: 162607-15-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...