2'-Hydroxy-4'-methylacetophenone ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-(2-Hydroxy-4-methylphenyl)ethan-1-one, 1-(2-Hydroxy-4-methylphenyl)ethanone

H1530-25G H1530-5G
TCH1530-25GEA 265 USD
TCH1530-25G TCH1530-5G
2'-Hydroxy-4'-methylacetophenone ≥95.0% (by GC)
2'-Hydroxy-4'-methylacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 245°C
MDL Number: MFCD00100559
CAS Number: 6921-64-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR