Desyl bromide ≥95.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Bromo-2-phenylacetophenone
B4471-25G
B4471-5G
TCB4471-25GEA
413
USD
TCB4471-25G
TCB4471-5G
Desyl bromide ≥95.0% (by GC)
Desyl bromide
Formula:
C₁₄H₁₁BrO MW: 275.14 g/mol |
MDL Number:
MFCD00000136 CAS Number: 1484-50-0 EINECS: 216-057-1 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...