2-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Fluoropyridine-3-boronic acid pinacol ester, 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

F1105-1G F1105-5G
TCF1105-1GEA 53 USD
TCF1105-1G TCF1105-5G
2-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Fluoro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MW: 223.05 g/mol
MDL Number: MFCD06798245
CAS Number: 452972-14-4

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR