2,6-Difluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,6-Difluoropyridine-3-boronic acid, pinacol ester, 2,6-Difluoropyridine-3-boronic acid Pinacol ester, 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2,6-Difluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
D5142-1G
D5142-200MG
76219-180EA
92
USD
76219-180
76219-182
2,6-Difluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2,6-Difluoro-3-pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₄BF₂NO₂ MW: 241.04 |
MDL Number:
MFCD09037475 CAS Number: 1072945-00-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...