6-Bromo-2-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

B5077-1G B5077-5G
TCB5077-1GEA 53 USD
TCB5077-1G TCB5077-5G
6-Bromo-2-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
6-Bromo-2-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BBrNO₂
MDL Number: MFCD06796177
CAS Number: 651358-83-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR