5-Cyano-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3514-1G T3514-200MG
TCT3514-1GEA 133 USD
TCT3514-1G TCT3514-200MG
5-Cyano-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
5-Cyano-3-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₅BN₂O₂
MDL Number: MFCD07780755
CAS Number: 402718-29-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR