4-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester
T3196-25G
T3196-5G
TCT3196-25GEA
162
USD
TCT3196-25G
TCT3196-5G
4-Methoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Methoxyphenylboronic acid pinacol ester
Formula:
C₁₃H₁₉BO₃ MW: 234.1 g/mol |
MDL Number:
MFCD05663864 CAS Number: 171364-79-7 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...