2,6-Difluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
D5006-1G
D5006-5G
76219-128EA
54
USD
76219-128
76219-130
2,6-Difluoro-3-methoxyphenylboronic acid (contains varying amounts of Anhydride)
2,6-Difluoro-3-methoxyphenylboronic acid
Formula:
C₇H₇BF₂O₃ MW: 187.94 g/mol |
MDL Number:
MFCD06657880 CAS Number: 870779-02-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...