2,6-Difluoro-3-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1210-1G E1210-5G
TCE1210-1GEA 65 USD
TCE1210-1G TCE1210-5G
2,6-Difluoro-3-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2,6-Difluoro-3-ethoxyphenylboronic acid
Formula: C₈H₉BF₂O₃
MDL Number: MFCD07369735
CAS Number: 849062-00-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR