2,3-Difluoro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
2,3-Difluoro-4-ethoxybenzeneboronic acid, 4-Ethoxy-2,3-difluorobenzeneboronic acid, 4-Ethoxy-2,3-difluorophenylboronic acid, (4-Ethoxy-2,3-difluorophenyl)boronic acid
E1192-1G
E1192-5G
TCE1192-1GEA
32
USD
TCE1192-1G
TCE1192-5G
2,3-Difluoro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2,3-Difluoro-4-ethoxyphenylboronic acid
Formula:
C₈H₉BF₂O₃ MW: 201.96 g/mol |
MDL Number:
MFCD09258743 CAS Number: 212386-71-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...