2-Fluoro-5-iodobenzotrifluoride ≥98.0% (by GC)

Supplier: TCI America
Warning

F1161-25G F1161-5G
TCF1161-25GEA 227 USD
TCF1161-25G TCF1161-5G
2-Fluoro-5-iodobenzotrifluoride ≥98.0% (by GC)
2-Fluoro-5-iodobenzotrifluoride
Formula: C₇H₃F₄I
MDL Number: MFCD00236678
CAS Number: 59382-39-7

Order Now

Specification Test Results

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR