3,5-Difluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D5017-1G D5017-5G
76219-772EA 67 USD
76219-772 76219-774
3,5-Difluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Difluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₂
MDL Number: MFCD11504963
CAS Number: 863868-36-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR