2-Fluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 2-Fluorophenylboronic acid, pinacol ester, 2-Fluorophenylboronic acid Pinacol ester

F1085-25G F1085-5G
76220-086EA 330 USD
76220-086 76220-088
2-Fluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Fluorophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BFO₂
MDL Number: MFCD05663874
CAS Number: 876062-39-4

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR