4-Bromophenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromophenylboronic acid, pinacol ester, 2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Bromophenylboronic acid Pinacol ester, 1-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

B4771-1G B4771-5G
TCB4771-1GEA 62 USD
TCB4771-1G TCB4771-5G
4-Bromophenylboronic acid pinacol ester ≥98.0% (by GC)
4-Bromophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BBrO₂
MDL Number: MFCD11110553
CAS Number: 68716-49-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR