3,5-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dimethoxyphenylboronic acid Pinacol ester

D5360-1G D5360-5G
76219-950EA 61 USD
76219-950 76219-952
3,5-Dimethoxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3,5-Dimethoxyphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₄
MW: 264.13 g/mol
MDL Number: MFCD05865191
CAS Number: 365564-07-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR