4-Bromo-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

B4691-1G B4691-5G
TCB4691-1GEA 83 USD
TCB4691-1G TCB4691-5G
4-Bromo-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Bromo-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MDL Number: MFCD03095347
CAS Number: 1735-53-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR